missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol, 99.5%, unstab., Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 04452622

Catalog No. AA4452622


Explore available promotions

Description

Description

As a precursor to many materials and useful compounds. Its major uses involve its conversion to plastics or related materials. Phenol and its chemical derivatives are key for building polycarbonates, epoxies, Bakelite, nylon, detergents, and numerous pharmaceutical drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenol
108-95-2
1.071 g/mL
79°C (175°F)
C6H6O
100 g
969616
14,7241
Moderately soluble in water (8.4% @ 20°C),alcohol,ether,chloroform,ethyl acetate,toluene,and glycerol.
OC1=CC=CC=C1
94.11
CHEBI:15882
99.5%
UnStabilized
40°C to 43°C
181°C
Sweet
MFCD00002143
UN1671
Hygroscopic
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
94.11
Solid Melt

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol
C6H6O
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
OC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311+H331-H314-H335-H341-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID

EINECSNumber : 203-632-7

RTECSNumber : SJ3325000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only