Learn More
Phenol, 99+%, for biochemistry, loose crystals, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 180781000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenol | |
108-95-2 | |
Colorless to White | |
182.0°C | |
Authentic | |
Glass bottle | |
C6H5OH | |
100 g | |
0.05% max. | |
1.07 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
CHEBI:15882 | |
99+% | |
Loose Crystals |
99+% | |
39.0°C to 42.0°C | |
1.0700g/mL | |
79°C | |
99% min. (GC) | |
C6H6O | |
MFCD00002143 | |
06, 110 | |
01,828; 13,166; 14,88 | |
15, 7354 | |
Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether | |
OC1=CC=CC=C1 | |
94.11 | |
3.437 mPa.s (50°C) | |
94.11 | |
Biochemistry |
Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
996 | |
phenol |
C6H6O | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 203-632-7
RTECSNumber : SJ3325000
TSCA : TSCA
RUO – Research Use Only