missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol, 99%, extra pure, Thermo Scientific Chemicals

$60.79 - $255.22

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149340500
SDS View Documents
Thermo Scientific Chemicals
149340500
50 g Glass bottle
Each for $60.79
 
AC149340010
SDS View Documents
Thermo Scientific Chemicals
149340010
1 kg Glass bottle
Each for $255.22
Case of 6 Each for $0.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
996
phenol
C6H6O
MFCD00002143
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
CHEBI:15882
OC1=CC=CC=C1

Specifications

108-95-2
Colorless
182.0°C
Authentic
Glass bottle
C6H5OH
50 g
01,828; 13,166; 14,88
15, 7354
Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
OC1=CC=CC=C1
94.11
3.437 mPa.s (50°C)
94.11
99%
Crystals or Fused Mass
39.0°C to 42.0°C
1.0700g/mL
79°C
98.5% min. (GC)
C6H6O
MFCD00002143
06, 110
1.07
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
CHEBI:15882
0.4mbar at 20°C
Extra Pure
Phenol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Danger

EINECSNumber : 203-632-7

Recommended Storage : Turns pink on exposure to air

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only