missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenol, Fused Crystal, Technical, Spectrum™ Chemical
$410.18 - $21816.82
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18-612-223
|
Spectrum Chemical Mfg Cor
P1065500GM |
500 g | Fiber Drum |
Each for $410.18
|
|
||||
18-612-222
|
Spectrum Chemical Mfg Cor
P10651KG |
1 kg | Fiber Drum |
Each for $584.88
|
|
||||
18-612-221
|
Spectrum Chemical Mfg Cor
P1065180KGBL |
180 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
108-95-2 |
Specifications
108-95-2 | |
182°C | |
500 g | |
Crystalline Powder or Lumps |
1 | |
Fiber Drum | |
Technical |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.