Learn More
Phenol (Liquid/Certified ACS), Fisher Chemical™
$1000.50 - $1565.00
Chemical Identifiers
| CAS | 108-95-2 |
|---|---|
| Molecular Formula | C6H6O |
| Molecular Weight (g/mol) | 94.11 |
| MDL Number | MFCD00002143 |
| InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
| Synonym | carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol |
| PubChem CID | 996 |
| ChEBI | CHEBI:15882 |
| IUPAC Name | phenol |
| SMILES | OC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
A931I-500
|
Thermo Fisher Scientific
FLA931I500 |
500 mL | Glass Bottle |
Each for $1,000.50
|
|
||||
|
A931I-1
|
Fisher Chemical
A931I1 |
1 L | Glass Bottle |
Each for $1,450.67
|
|
||||
|
A931I-4
|
Fisher Chemical
A931I4 |
4 L | Glass Bottle |
Each for $1,565.00
|
|
||||
Chemical Identifiers
| 108-95-2 | |
| 94.11 | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| 996 | |
| phenol |
| C6H6O | |
| MFCD00002143 | |
| carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
| CHEBI:15882 | |
| OC1=CC=CC=C1 |
Specifications
| 108-95-2 | |
| Colorless | |
| 6 | |
| 79.4°C | |
| C6H6O | |
| 500 mL | |
| 0.05% max. | |
| ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
| phenol | |
| 996 | |
| 0.35mmHg at 25°C | |
| Certified | |
| Phenol |
| 42.8°C | |
| 1.0576g/cm³ | |
| 182°C | |
| Glass Bottle | |
| MFCD00002143 | |
| 2821 | |
| carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
| OC1=CC=CC=C1 | |
| 94.11 | |
| CHEBI:15882 | |
| ≥89% | |
| Liquid |
Safety and Handling
DANGER!
Emergency Overview
Combustible liquid. Toxic by inhalation. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:4
Flammability:2
Instability:0
DOTInformation : DOT Class 6.1, : Poison