missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Phenol, Molecular Biology Grade, Calbiochem™,
Supplier: MilliporeSigma™ 516724500GM
Specifications
Phenol | |
40.8°C | |
1.06g/cm3 (20°C) | |
181.8°C | |
Molecular biology grade | |
C6H6O | |
500 g | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
94.1 | |
≥99.0% | |
Solid |
108-95-2 | |
Colorless | |
5 (at 50g/L 20°C) | |
595°C (1103°F) | |
Characteristic | |
MFCD00002143 | |
UN 1671 | |
Water: 84g/L at 20°C soluble | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
0.2 hPa (20°C) | |
GC |
Chemical Identifiers
108-95-2 | |
Colorless | |
40.8°C |
C6H6O | |
UN 1671 | |
Phenol |
Safety and Handling
H314: Causes severe skin burns and eye damage.
H341: Suspected of causing genetic defects.
H301-H311-H331: Toxic if swallowed. In contact with skin or if inhaled
DOTInformation : DOT Class: 6.1, Packaging Group: II
RTECSNumber : SJ3325000
Recommended Storage : -20°C (-4°F), Tightly closed. Dry. Keep in a well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized person.