missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenol Red Indicator, ACS Reagent, Honeywell Fluka™
Indicator, ACS reagent
Supplier: Honeywell-Fluka 32661100G
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Phenol Red | |
C19H14O5S | |
NONH for all modes of transport | |
Phenolsulfonphthalein | |
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
354.38 | |
CHEBI:31991 | |
ACS Reagent | |
6.8 to 8.2 (yellow to red) | |
100 g |
143-74-8 | |
MFCD00003552 | |
326470 | |
BELBBZDIHDAJOR-UHFFFAOYSA-N | |
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
4766 | |
354.38g/mol | |
Glass Bottle | |
Red | |
Powder |
Chemical Identifiers
143-74-8 | |
354.38 | |
BELBBZDIHDAJOR-UHFFFAOYSA-N | |
4766 | |
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
C19H14O5S | |
MFCD00003552 | |
Phenolsulfonphthalein | |
CHEBI:31991 | |
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Safety and Handling
P261-P305 + P351 + P338
H315-H319-H335
EINECSNumber : 205-609-7
RTECSNumber : SJ7490000