missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol Standard, Certified, 1000ppm ±10ppm (1mL = 1mg), LabChem™

Phenol Standard, C6H6O, CAS Number-108-95-2, 7732-18-5, 500mL, 99.9, 0.1, CHEBI:15882, Colorless, 1000ppm +/-10ppm (1mL - 1mg), 1g/mL, 94.11, Certified, GHS H Statement Solution is not hazardous.

Supplier:  LabChem LC183301

 View more versions of this product

Catalog No. LC183301


Explore available promotions

Description

Description

Phenol Standard, C6H6O, CAS Number-108-95-2, 7732-18-5, 500mL, 99.9, 0.1, CHEBI:15882, Colorless, 1000ppm +/-10ppm (1mL - 1mg), 1g/mL, 94.11, Certified, GHS H Statement

Solution is not hazardous.

Specifications

Specifications

Phenol Standard
1000ppm (1mL = 1mg)
Amber Glass
99.9,0.1
Colorless
C6H5O8
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
94.11
Passes Test
Soluble in water
1000ppm ±10ppm (1mL = 1mg)
108-95-2,7732-18-5
500 mL
Buffer Solution
C6H6O
MFCD00002143
OC1=CC=CC=C1
94.11
CHEBI:15882
Certified
1g/mL
Liquid
Videos
SDS
Documents

Documents

Promotions

Promotions