Learn More
Thermo Scientific Chemicals Phenol, ultrapure, 99%, unstab.
Supplier: Thermo Scientific Chemicals J6401136
Description
Phenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenol can be used for a wide variety of molecular biology applications, including nucleic acid extractions using standard phenol-chloroform protocols. Epidermal strips from the abaxial side of leaves of Commelina communis were stirred in phenol in a procedure to separate the monosaccharides from the epidermis.
Solubility
Soluble in water (82 g/L at 20°C).
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications
Phenol | |
108-95-2 | |
White | |
181°C | |
Ultrapure | |
C6H6O | |
500 g | |
969616 | |
14,7241 | |
Soluble in water (82g/L at 20°C). | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
99% |
unstabilized | |
40°C to 43°C | |
1.071 | |
79°C (175°F) | |
Sweet | |
MFCD00002143 | |
UN1671 | |
Air Sensitive | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
94.11 | |
Crystalline |
Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
996 | |
phenol |
C6H6O | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H301-H311-H331-H314-H341-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335-H341-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: PHENOL, SOLID
EINECSNumber : 203-632-7
RTECSNumber : SJ3325000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only