Fisherbrand™ Phenolic Shelving
Fisherbrand™ Phenolic Shelving is made out of chemical resistant black phenolic resin. To be used with wall standards or island stanchions.
 View more versions of this product
Catalog No. 06000082
Explore available promotions
Description
Description
  • 1 in. thick black phenolic resin shelf
  • Front Edge is finished
Specifications
Specifications
Shelf
Phenolic
20.3 cm
2.5 cm
91.4 cm
Black
8 in.
1 in.
36 in.