Fisherbrand™ Phenolic Wall Cabinet, 2 Framed Glass Doors
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
Description
- Rust, rot, mold resistant construction.
- Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
- Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
- All colors have a solid black core
Specifications
2 years | |
13 in. | |
30 in. | |
Framed Glass Door | |
91.4 cm | |
2 | |
Phenolic |
2 | |
76.2 cm | |
36 in. | |
33 cm | |
Wall | |
Beige |