missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenolphthalein (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLP79100
Specifications
Phenolphthalein | |
77-09-8 | |
C20H14O4 | |
MFCD00005913 | |
KJFMBFZCATUALV-UHFFFAOYSA-N | |
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one | |
4764 | |
318.33 | |
Glass Bottle | |
Pass Test | |
100 g |
258°C | |
Pass Test | |
C6H4COOC(C6H4-4-OH)2 | |
3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone, 3, 3-Bis(p-hydroxyphenyl) Phthallide | |
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
318.33 | |
CHEBI:34914 | |
Certified ACS | |
Vapor Pressure: Negligible | |
White | |
Solid |
Chemical Identifiers
77-09-8 | |
318.33 | |
KJFMBFZCATUALV-UHFFFAOYSA-N | |
4764 | |
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
C20H14O4 | |
MFCD00005913 | |
3, 3-Bis(p-hydroxyphenyl)-1(3H)-isobenzofuranone, 3, 3-Bis(p-hydroxyphenyl) Phthallide | |
CHEBI:34914 | |
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |