missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenoxazine, 97%, Thermo Scientific Chemicals
$93.41 - $93.41
Chemical Identifiers
CAS | 135-67-1 |
---|---|
Molecular Formula | C12H9NO |
Molecular Weight (g/mol) | 183.21 |
MDL Number | MFCD00005014 |
InChI Key | TZMSYXZUNZXBOL-UHFFFAOYSA-N |
Synonym | phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci |
PubChem CID | 67278 |
IUPAC Name | 10H-phenoxazine |
SMILES | N1C2=CC=CC=C2OC2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC130220010
|
Thermo Scientific Chemicals
130220010 |
1 g | Glass bottle |
Each for $93.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
135-67-1 | |
183.21 | |
TZMSYXZUNZXBOL-UHFFFAOYSA-N | |
67278 | |
N1C2=CC=CC=C2OC2=CC=CC=C12 |
C12H9NO | |
MFCD00005014 | |
phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci | |
10H-phenoxazine |
Specifications
135-67-1 | |
Gray to Gray-Green | |
96% min. (HPLC) | |
C12H9NO | |
1 g | |
15, 7365 | |
Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether | |
N1C2=CC=CC=C2OC2=CC=CC=C12 | |
183.21 | |
183.21 | |
Fine Crystalline Powder |
154.0°C to 159.0°C | |
Authentic | |
Glass bottle | |
MFCD00005014 | |
27, 62 | |
phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci | |
TZMSYXZUNZXBOL-UHFFFAOYSA-N | |
10H-phenoxazine | |
67278 | |
97% | |
Phenoxazine |
Safety and Handling
EINECSNumber : 205-210-8
RUO – Research Use Only