missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxyacetone 97.0+%, TCI America™
$242.33 - $842.56
Chemical Identifiers
| CAS | 621-87-4 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00008767 |
| InChI Key | QWAVNXZAQASOML-UHFFFAOYSA-N |
| Synonym | phenoxyacetone, 2-propanone, 1-phenoxy, 1-phenoxy-2-propanone, 1-phenoxyacetone, 1-phenoxy-propan-2-one, phenoxy-2-propanone, phenoxymethyl methyl ketone, phenoxyaceton, phenoxy acetone, pubchem20265 |
| PubChem CID | 69313 |
| IUPAC Name | 1-phenoxypropan-2-one |
| SMILES | CC(=O)COC1=CC=CC=C1 |
Chemical Identifiers
| 621-87-4 | |
| 150.18 | |
| QWAVNXZAQASOML-UHFFFAOYSA-N | |
| 69313 | |
| CC(=O)COC1=CC=CC=C1 |
| C9H10O2 | |
| MFCD00008767 | |
| phenoxyacetone, 2-propanone, 1-phenoxy, 1-phenoxy-2-propanone, 1-phenoxyacetone, 1-phenoxy-propan-2-one, phenoxy-2-propanone, phenoxymethyl methyl ketone, phenoxyaceton, phenoxy acetone, pubchem20265 | |
| 1-phenoxypropan-2-one |
Specifications
| 621-87-4 | |
| 230°C | |
| MFCD00008767 | |
| phenoxyacetone, 2-propanone, 1-phenoxy, 1-phenoxy-2-propanone, 1-phenoxyacetone, 1-phenoxy-propan-2-one, phenoxy-2-propanone, phenoxymethyl methyl ketone, phenoxyaceton, phenoxy acetone, pubchem20265 | |
| CC(=O)COC1=CC=CC=C1 | |
| 150.18 | |
| 150.18 | |
| Liquid |
| Yellow | |
| C9H10O2 | |
| 5 g | |
| QWAVNXZAQASOML-UHFFFAOYSA-N | |
| 1-phenoxypropan-2-one | |
| 69313 | |
| ≥97.0% (GC) | |
| Phenoxyacetone |
Safety and Handling
TSCA : Yes