missing translation for 'onlineSavingsMsg'
Learn More

Phenoxydiphenylphosphine 98.0+%, TCI America™

$257.24 - $1,133.35

Chemical Identifiers

CAS 13360-92-4
Molecular Formula C18H15OP
Molecular Weight (g/mol) 278.291
InChI Key UPDNYUVJHQABBS-UHFFFAOYSA-N
Synonym Phenyl Diphenylphosphinite
PubChem CID 11543680
IUPAC Name phenoxy(diphenyl)phosphane
SMILES C1=CC=C(C=C1)OP(C2=CC=CC=C2)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P18435G
View Documents
TCI America
P18435G
5 g N/A
Only null left
 
P184325G
View Documents
TCI America
P184325G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13360-92-4
278.291
Phenyl Diphenylphosphinite
phenoxy(diphenyl)phosphane
C18H15OP
UPDNYUVJHQABBS-UHFFFAOYSA-N
11543680
C1=CC=C(C=C1)OP(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

13360-92-4
170°C
5 g
UPDNYUVJHQABBS-UHFFFAOYSA-N
phenoxy(diphenyl)phosphane
11543680
≥98.0% (GC)
Phenoxydiphenylphosphine
Yellow
C18H15OP
Phenyl Diphenylphosphinite
C1=CC=C(C=C1)OP(C2=CC=CC=C2)C3=CC=CC=C3
278.291
278.29
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'