missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl 1-Propenyl Ketone 80.0+%, TCI America™
$615.14 - $922.71
Chemical Identifiers
| CAS | 495-41-0 |
|---|---|
| Molecular Formula | C10H10O |
| Molecular Weight (g/mol) | 146.189 |
| MDL Number | MFCD00059386 |
| InChI Key | FUJZJBCWPIOHHN-QHHAFSJGSA-N |
| Synonym | 1-Phenyl-2-buten-1-one |
| PubChem CID | 231934 |
| IUPAC Name | (E)-1-phenylbut-2-en-1-one |
| SMILES | CC=CC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 495-41-0 | |
| 146.189 | |
| FUJZJBCWPIOHHN-QHHAFSJGSA-N | |
| 231934 | |
| CC=CC(=O)C1=CC=CC=C1 |
| C10H10O | |
| MFCD00059386 | |
| 1-Phenyl-2-buten-1-one | |
| (E)-1-phenylbut-2-en-1-one |
Specifications
| 495-41-0 | |
| 122°C | |
| MFCD00059386 | |
| 1-Phenyl-2-buten-1-one | |
| CC=CC(=O)C1=CC=CC=C1 | |
| 146.189 | |
| 146.19 | |
| Liquid |
| Yellow | |
| C10H10O | |
| 10 g | |
| FUJZJBCWPIOHHN-QHHAFSJGSA-N | |
| (E)-1-phenylbut-2-en-1-one | |
| 231934 | |
| ≥80.0% (GC) | |
| Phenyl 1-Propenyl Ketone |
Safety and Handling
TSCA : Yes