missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-galactopyranoside 98.0+%, TCI America™
Supplier: TCI America P16425G
Specifications
| Phenyl 3,4,6-Tri-O-acetyl-2-deoxy-1-thio-2-(2,2,2-trichloroethoxyformamido)-beta-D-galactopyranoside | |
| 117°C | |
| C21H24Cl3NO9S | |
| 5 g | |
| CC(=O)OCC1OC(SC2=CC=CC=C2)C(NC(=O)OCC(Cl)(Cl)Cl)C(OC(C)=O)C1OC(C)=O | |
| 572.83 | |
| 572.83 | |
| Crystalline Powder |
| 278784-83-1 | |
| White | |
| MFCD11112181,MFCD15072189 | |
| AZDNCEYOFMNMSH-UHFFFAOYNA-N | |
| [3,4-bis(acetyloxy)-6-(phenylsulfanyl)-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}oxan-2-yl]methyl acetate | |
| 133554192 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 278784-83-1 | |
| 572.83 | |
| AZDNCEYOFMNMSH-UHFFFAOYNA-N | |
| [3,4-bis(acetyloxy)-6-(phenylsulfanyl)-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}oxan-2-yl]methyl acetate |
| C21H24Cl3NO9S | |
| MFCD11112181,MFCD15072189 | |
| 133554192 | |
| CC(=O)OCC1OC(SC2=CC=CC=C2)C(NC(=O)OCC(Cl)(Cl)Cl)C(OC(C)=O)C1OC(C)=O |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator