Learn More
Phenyl acetate, 97%
CAS: 122-79-2 | C8H8O2 | 136.15 g/mol
$41.81 - $80.71
Identifiants chimiques
| CAS | 122-79-2 |
|---|---|
| Molecular Formula | C8H8O2 |
| Molecular Weight (g/mol) | 136.15 |
| MDL Number | MFCD00008699 |
| InChI Key | IPBVNPXQWQGGJP-UHFFFAOYSA-N |
| Synonym | acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech |
| PubChem CID | 31229 |
| ChEBI | CHEBI:8082 |
| IUPAC Name | phenyl acetate |
| SMILES | CC(=O)OC1=CC=CC=C1 |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC148770050
|
Thermo Scientific Chemicals
148770050 |
5 g | Glass bottle |
chaque for $41.81
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC148771000
|
Thermo Scientific Chemicals
148771000 |
100 g | Glass bottle |
chaque for $80.71
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 122-79-2 | |
| 136.15 | |
| IPBVNPXQWQGGJP-UHFFFAOYSA-N | |
| 31229 | |
| phenyl acetate |
| C8H8O2 | |
| MFCD00008699 | |
| acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech | |
| CHEBI:8082 | |
| CC(=O)OC1=CC=CC=C1 |
Spécifications
| 122-79-2 | |
| 1.0700g/mL | |
| 76°C | |
| 96% min. (GC) | |
| C8H8O2 | |
| CH3CO2C6H5 | |
| 5 g | |
| 01,829 | |
| 15,7379 | |
| Solubility in water: ca 4g/L (25°C). Other solubilities: miscible with alcohol,chloroform,ether,soluble in glacial acetic acid | |
| CC(=O)OC1=CC=CC=C1 | |
| 136.15 | |
| CHEBI:8082 | |
| 97% | |
| Phenyl acetate, 97% |
| Colorless to Brown | |
| 196.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5010 to 1.5030 | |
| MFCD00008699 | |
| 06,152 | |
| 1.07 | |
| acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech | |
| IPBVNPXQWQGGJP-UHFFFAOYSA-N | |
| phenyl acetate | |
| 31229 | |
| 136.15 | |
| Liquid |
Sécurité et manipulation
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-575-0
missing translation for 'rtecsNumber' : AJ2800000
missing translation for 'tsca' : TSCA
RUO – Research Use Only