Learn More
Phenyl acetate, 97%, Thermo Scientific Chemicals
$85.75 - $241.64
Chemical Identifiers
CAS | 122-79-2 |
---|---|
Molecular Formula | C8H8O2 |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00008699 |
InChI Key | IPBVNPXQWQGGJP-UHFFFAOYSA-N |
Synonym | acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech |
PubChem CID | 31229 |
ChEBI | CHEBI:8082 |
IUPAC Name | phenyl acetate |
SMILES | CC(=O)OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2256130
|
Thermo Scientific Chemicals
B2256130 |
250 g |
Each for $85.75
|
|
|||||
AAB225610B
|
Thermo Scientific Chemicals
B225610B |
1000 g |
Each for $241.64
|
|
|||||
Description
Phenyl acetate levels in urine are marker for the diagnosis of some forms of unipolar major depressive disorders. It undergoes Fries rearrangement to form a mixture of o- and p-hydroxyacetophenones which are useful intermediates in manufacture of pharmaceuticals. It is a metabolite of Phenylbutyrate (PB), useful in the treatment of neuroblastoma and lung cancer. An odorant found in strawberries, passion fruit and black tea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (4 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
122-79-2 | |
136.15 | |
IPBVNPXQWQGGJP-UHFFFAOYSA-N | |
31229 | |
phenyl acetate |
C8H8O2 | |
MFCD00008699 | |
acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech | |
CHEBI:8082 | |
CC(=O)OC1=CC=CC=C1 |
Specifications
122-79-2 | |
195°C to 196°C | |
Phenol-like | |
1.502 | |
250 g | |
14,7267 | |
Soluble in water (4g/L at 20°C). | |
CC(=O)OC1=CC=CC=C1 | |
136.15 | |
CHEBI:8082 | |
97% |
1.075 | |
76°C (168°F) | |
C8H8O2 | |
MFCD00008699 | |
636458 | |
acetic acid phenyl ester, phenol acetate, acetylphenol, acetic acid, phenyl ester, acetyl phenol, acetoxybenzene, acetates, acetic acid,phenyl ester, fenylester kyseliny octove, fenylester kyseliny octove czech | |
IPBVNPXQWQGGJP-UHFFFAOYSA-N | |
phenyl acetate | |
31229 | |
136.15 | |
Phenyl acetate |
Safety and Handling
GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
EINECSNumber : 204-575-0
RTECSNumber : AJ2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only