Learn More
Phenyl acrylate, 97%, Thermo Scientific Chemicals
$147.18 - $544.56
Chemical Identifiers
CAS | 937-41-7 |
---|---|
Molecular Formula | C9H8O2 |
Molecular Weight (g/mol) | 148.161 |
MDL Number | MFCD00048145 |
InChI Key | WRAQQYDMVSCOTE-UHFFFAOYSA-N |
Synonym | phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht |
PubChem CID | 61242 |
IUPAC Name | phenyl prop-2-enoate |
SMILES | C=CC(=O)OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1181906
|
Thermo Scientific Chemicals
L1181906 |
5 g |
Each for $147.18
|
|
|||||
AAL1181914
|
Thermo Scientific Chemicals
L1181914 |
25 g |
Each for $544.56
|
|
|||||
Description
Phenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl acrylate reacts with benzaldehyde to produce 2-(hydroxy-phenyl-methyl)-acrylic acid phenyl ester.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store at 4°C. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
937-41-7 | |
148.161 | |
WRAQQYDMVSCOTE-UHFFFAOYSA-N | |
61242 | |
C=CC(=O)OC1=CC=CC=C1 |
C9H8O2 | |
MFCD00048145 | |
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht | |
phenyl prop-2-enoate |
Specifications
937-41-7 | |
89°C to 90°C (12 mmHg) | |
1.521 | |
5 g | |
2041124 | |
Insoluble in water. | |
C=CC(=O)OC1=CC=CC=C1 | |
148.161 | |
148.16 | |
Phenyl acrylate |
1.076 | |
C9H8O2 | |
MFCD00048145 | |
UN3082 | |
phenyl acrylate, phenylacrylate, 2-propenoic acid, phenyl ester, phenol acrylate, phenyl-acrylate, acrylic acid phenyl ester, phenyl acrylate 5g, 2-propenoic acid,phenyl ester, acrylic acid phenyl ester, stabilized with bht | |
WRAQQYDMVSCOTE-UHFFFAOYSA-N | |
phenyl prop-2-enoate | |
61242 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 213-329-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only