missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl beta-D-Glucopyranoside Hydrate 99.0+%, TCI America™
$37.29 - $37.29
Chemical Identifiers
| CAS | 1464-44-4 |
|---|---|
| Molecular Formula | C12H16O6 |
| Molecular Weight (g/mol) | 256.25 |
| MDL Number | MFCD03410292 |
| InChI Key | NEZJDVYDSZTRFS-WLEIBRHLNA-N |
| Synonym | phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d |
| PubChem CID | 65080 |
| ChEBI | CHEBI:28749 |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol |
| SMILES | OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O |
Chemical Identifiers
| 1464-44-4 | |
| 256.25 | |
| NEZJDVYDSZTRFS-WLEIBRHLNA-N | |
| 65080 | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol |
| C12H16O6 | |
| MFCD03410292 | |
| phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d | |
| CHEBI:28749 | |
| OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O |
Specifications
| 1464-44-4 | |
| White | |
| MFCD03410292 | |
| phenyl beta-d-glucopyranoside, phenylglucoside, phenyl-beta-d-glucopyranoside, phenol glucoside, phenyl beta-d-glucoside, phenyl-beta-d-glucoside, .beta.-d-glucopyranoside, phenyl, unii-4w3pgi3766, aryl beta-d-glucoside, glucopyranoside, phenyl-, beta-d | |
| OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O | |
| 256.25 | |
| CHEBI:28749 | |
| ≥99.0% (GC) | |
| Phenyl beta-D-Glucopyranoside Hydrate |
| 173°C | |
| C12H16O6 | |
| 1 g | |
| NEZJDVYDSZTRFS-WLEIBRHLNA-N | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol | |
| 65080 | |
| 256.25 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : LZ5985510
TSCA : Yes