missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl bromoacetate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6046418

 View more versions of this product

Catalog No. AAH6046418


Explore available promotions

Description

Description

Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl bromoacetate may be employed as alkylation reagent in the preparation of 2-(phenoxycarbonyl)methyl triazoles. It may be used in the synthesis of the A-ring of cylindrospermopsin and in the synthesis of 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3, 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-3-methyl-1,3-thiazolidin-4-one3, 3-ethyl-2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene]hydrazinyl}-1,3-thiazolidin-4-one3 and 2-{2-[1-(4-hydroxy-6-methyl-2-oxo-2H-pyran-3-yl)ethylidene] hydrazinyl}-3-phenyl-1,3-thiazolidin-4-one3.

Solubility
Soluble in ethanol and ether, insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.
Specifications

Specifications

Phenyl bromoacetate
31°C to 33°C
134°C
C8H7BrO2
MFCD00192391
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
UEWYUCGVQMZMGY-UHFFFAOYSA-N
phenyl 2-bromoacetate
564919
98%
620-72-4
1.508
113°C (235°F)
BrCH2CO2C6H5
50 g
Soluble in ethanol and ether,insoluble in water.
C1=CC=C(C=C1)OC(=O)CBr
215.046
215.04

Chemical Identifiers

620-72-4
215.046
UEWYUCGVQMZMGY-UHFFFAOYSA-N
564919
C1=CC=C(C=C1)OC(=O)CBr
C8H7BrO2
MFCD00192391
phenyl bromoacetate, acetic acid, bromo-, phenyl ester, bromoacetic acid phenyl ester, bromoacetic acid, phenyl ester, phenylbromessigester, phenyl bromoacetate #, acmc-209mzb, phenyl 2-bromanylethanoate, ksc352o8b
phenyl 2-bromoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only