missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenyl chloroformate, 99%, Thermo Scientific Chemicals
$171.57 - $171.57
Chemical Identifiers
CAS | 1885-14-9 |
---|---|
Molecular Formula | C7H5ClO2 |
Molecular Weight (g/mol) | 156.57 |
MDL Number | MFCD00000637 |
InChI Key | AHWALFGBDFAJAI-UHFFFAOYSA-N |
Synonym | phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y |
PubChem CID | 15891 |
IUPAC Name | phenyl carbonochloridate |
SMILES | ClC(=O)OC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC218212500
|
Thermo Scientific Chemicals
218212500 |
250 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
1885-14-9 | |
156.57 | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
15891 | |
ClC(=O)OC1=CC=CC=C1 |
C7H5ClO2 | |
MFCD00000637 | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
phenyl carbonochloridate |
Specifications
1885-14-9 | |
1.2440g/mL | |
80°C | |
98.5% min. (GC) | |
C7H5ClO2 | |
ClCO2C6H5 | |
250 mL | |
phenyl chloroformate, chloroformic acid phenyl ester, phenyl chlorocarbonate, carbonochloridic acid, phenyl ester, phenoxycarbonyl chloride, formic acid, chloro-, phenyl ester, fenylester kyseliny chlormravenci, phenylchloroformate, unii-6tnd0d6d3y, 6tnd0d6d3y | |
AHWALFGBDFAJAI-UHFFFAOYSA-N | |
phenyl carbonochloridate | |
15891 | |
156.57 | |
Liquid |
-28.0°C | |
188.0°C to 189.0°C (1013.0 mbar) | |
Authentic | |
Glass bottle | |
1.5100 to 1.5120 | |
MFCD00000637 | |
1.244 | |
Solubility in water: hydrolysis | |
ClC(=O)OC1=CC=CC=C1 | |
156.57 | |
1.86 mPa.s (20°C) | |
99% | |
Phenyl chloroformate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 217-547-8
RUO – Research Use Only