missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl ether, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130601000

 View more versions of this product

Catalog No. AC130601000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenyl ether
26.0°C to 30.0°C
259.0°C
98.5% min. (GC)
C12H10O
(C6H5)2O
100 g
1.073
diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl
USIUVYZYUHIAEV-UHFFFAOYSA-N
phenoxybenzene
7583
CHEBI:39258
99%
101-84-8
1.0730g/mL
115°C
Glass bottle
1.5795 to 1.5815
MFCD00003034
06,146
15,3357
Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether and,glacial acetic acid
C1=CC=C(C=C1)OC2=CC=CC=C2
170.21
3.4909 mPa.s (28°C)
170.21

Chemical Identifiers

101-84-8
170.21
USIUVYZYUHIAEV-UHFFFAOYSA-N
7583
phenoxybenzene
C12H10O
MFCD00003034
diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl
CHEBI:39258
C1=CC=C(C=C1)OC2=CC=CC=C2
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN

GHS Signal Word: Warning

EINECSNumber : 202-981-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only