Learn More
Phenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1159622
Description
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.
Solubility
Soluble in alcohol, and ether. Insoluble in water.
Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Store at 4°C.
Specifications
Phenyl isothiocyanate | |
-21°C | |
220°C to 221°C | |
Pungent | |
1.65 | |
100 g | |
471392 | |
14,7297 | |
Soluble in alcohol,and ether. Insoluble in water. | |
S=C=NC1=CC=CC=C1 | |
135.18 | |
CHEBI:85103 | |
97% |
103-72-0 | |
1.131 | |
87°C (188°F) | |
C7H5NS | |
MFCD00004798 | |
UN2927 | |
Moisture sensitive | |
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate | |
QKFJKGMPGYROCL-UHFFFAOYSA-N | |
isothiocyanatobenzene | |
7673 | |
135.19 |
Chemical Identifiers
103-72-0 | |
135.18 | |
QKFJKGMPGYROCL-UHFFFAOYSA-N | |
7673 | |
isothiocyanatobenzene |
C7H5NS | |
MFCD00004798 | |
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate | |
CHEBI:85103 | |
S=C=NC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H301-H334-H314-H318-H227-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
H227-H301+H311+H331-H314-H317-H334-H335-H361d
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber : 203-138-1
RTECSNumber : NX9275000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only