missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1159636

 View more versions of this product

Catalog No. AAA1159636



Description

Description

Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of
Specifications

Specifications

Phenyl isothiocyanate
-21°C
220°C to 221°C
Pungent
1.65
500 g
471392
14,7297
Soluble in alcohol,and ether. Insoluble in water.
S=C=NC1=CC=CC=C1
135.18
CHEBI:85103
97%
103-72-0
1.131
87°C (188°F)
C7H5NS
MFCD00004798
UN2927
Moisture sensitive
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
QKFJKGMPGYROCL-UHFFFAOYSA-N
isothiocyanatobenzene
7673
135.19

Chemical Identifiers

103-72-0
135.18
QKFJKGMPGYROCL-UHFFFAOYSA-N
7673
isothiocyanatobenzene
C7H5NS
MFCD00004798
phenyl isothiocyanate, phenylisothiocyanate, thiocarbanil, phenyl mustard oil, benzene, isothiocyanato, pitc, isothiocyanic acid phenyl ester, benzene-1-isothiocyanate, phenylsenfoel, phenyl thioisocyanate
CHEBI:85103
S=C=NC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H314-H318-H227-H317;
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.

P201-P202-P210-P235-P260-P264b-P270-P271-P272-P280g-P281-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H227-H301+H311+H331-H314-H317-H334-H335-H361d

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 203-138-1

RTECSNumber : NX9275000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only