Learn More
Phenyl salicylate, 99%
Phenyl salicylate, 99%, C13H10O3, CAS Number-118-55-8, salol, salphenyl, phenyl salicylate, 2-phenoxycarbonylphenol, salicylic acid, phenyl ester, benzoic acid, 2-hydroxy-, phenyl ester, phenyl-2-hydroxybenzoate, musol, phenol salicylate, 2-hydroxybenzoic acid phenyl ester, 250g, 98.5% min. (GC) | CAS: 118-55-8 | C13H10O3 | 214.22 g/mol
$96.11 - $308.69
Chemical Identifiers
| CAS | 118-55-8 |
|---|---|
| Molecular Formula | C13H10O3 |
| Molecular Weight (g/mol) | 214.22 |
| InChI Key | ZQBAKBUEJOMQEX-UHFFFAOYSA-N |
| Synonym | phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate |
| PubChem CID | 8361 |
| ChEBI | CHEBI:34918 |
| IUPAC Name | phenyl 2-hydroxybenzoate |
| SMILES | C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC16508-2500
|
Thermo Scientific Chemicals
165082500 |
250 g | Glass bottle |
Each for $96.11
|
|
||||
|
AC165080010
|
Thermo Scientific Chemicals
165080010 |
1 kg | Glass bottle |
Each for $308.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 118-55-8 | |
| 214.22 | |
| phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate | |
| CHEBI:34918 | |
| C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O |
| C13H10O3 | |
| ZQBAKBUEJOMQEX-UHFFFAOYSA-N | |
| 8361 | |
| phenyl 2-hydroxybenzoate |
Specifications
| 118-55-8 | |
| White | |
| >110°C | |
| 98.5% min. (GC) | |
| C13H10O3 | |
| 250 g | |
| 15,742 | |
| Solubility in water: <0.1% (25°C). Other solubilities: 1g/1.5mL benzene- 1g/5mL amyl alcohol,53g/100g abs.ethanol-470g/100g ethylacetate (25 c),620g/100g methylethylketone-460g/100g toluene(25 c,soluble in chloroform,acetone,ether and oils,s | |
| C1=CC=C(C=C1)OC(=O)C2=CC=CC=C2O | |
| 214.22 | |
| CHEBI:34918 | |
| 99% | |
| Phenyl salicylate |
| 40.0°C to 44.0°C | |
| 172.0°C to 173.0°C (12.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 2-(HO)C6H4CO2C6H5 | |
| 10,76 | |
| phenyl salicylate, salol, phenol salicylate, salphenyl, musol, 2-phenoxycarbonylphenol, benzoic acid, 2-hydroxy-, phenyl ester, salicylic acid, phenyl ester, 2-hydroxybenzoic acid phenyl ester, phenyl-2-hydroxybenzoate | |
| ZQBAKBUEJOMQEX-UHFFFAOYSA-N | |
| phenyl 2-hydroxybenzoate | |
| 8361 | |
| 214.22 | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 204-259-2
RUO – Research Use Only