missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl sulfone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130962500

 View more versions of this product

Catalog No. AC130962500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenyl sulfone
127-63-9
White
Authentic
Plastic bottle
(C6H5)2SO2
250 g
15, 3372
Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water,soluble in hot alcohol and benzene
O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
218.27
CHEBI:78360
97%
97%
125.0°C to 129.0°C
379.0°C
97%
C12H10O2S
MFCD00007548
06, 300
diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
KZTYYGOKRVBIMI-UHFFFAOYSA-N
(benzenesulfonyl)benzene
31386
218.28
Crystalline Powder

Chemical Identifiers

127-63-9
218.27
KZTYYGOKRVBIMI-UHFFFAOYSA-N
31386
O=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
C12H10O2S
MFCD00007548
diphenyl sulfone, phenyl sulfone, sulfonyldibenzene, diphenylsulfone, sulfobenzide, diphenyl sulphone, benzene, 1,1'-sulfonylbis, sulfone, diphenyl, 1,1'-sulfonyldibenzene, phenyl sulphone
CHEBI:78360
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Warning

EINECSNumber : 204-853-1

RTECSNumber : SX2400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only