missing translation for 'onlineSavingsMsg'
Learn More

Phenyl Trifluoromethyl Sulfone 98.0+%, TCI America™

$126.76 - $410.10

Chemical Identifiers

CAS 426-58-4
Molecular Formula C7H5F3O2S
Molecular Weight (g/mol) 210.17
MDL Number MFCD00159083
InChI Key UPGBQYFXKAKWQC-UHFFFAOYSA-N
PubChem CID 555605
IUPAC Name trifluoromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P21951G
View Documents
TCI America
P21951G
1 g N/A
Only null left
 
P21955G
View Documents
TCI America
P21955G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

426-58-4
210.17
UPGBQYFXKAKWQC-UHFFFAOYSA-N
trifluoromethylsulfonylbenzene
C7H5F3O2S
MFCD00159083
555605
C1=CC=C(C=C1)S(=O)(=O)C(F)(F)F

Specifications

426-58-4
203°C
MFCD00159083
UPGBQYFXKAKWQC-UHFFFAOYSA-N
trifluoromethylsulfonylbenzene
555605
≥98.0% (GC)
Phenyl Trifluoromethyl Sulfone
Colorless
C7H5F3O2S
1 g
C1=CC=C(C=C1)S(=O)(=O)C(F)(F)F
210.17
210.17
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'