Learn More
Phenylacetaldehyde, 95%, Thermo Scientific Chemicals
$41.79 - $312.74
Chemical Identifiers
CAS | 122-78-1 |
---|---|
Molecular Formula | C8H8O |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD00006993 |
InChI Key | DTUQWGWMVIHBKE-UHFFFAOYSA-N |
Synonym | phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde |
PubChem CID | 998 |
ChEBI | CHEBI:16424 |
IUPAC Name | 2-phenylacetaldehyde |
SMILES | O=CCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1426314
|
Thermo Scientific Chemicals
A1426314 |
25 g |
Each for $41.79
|
|
|||||
AAA1426322
|
Thermo Scientific Chemicals
A1426322 |
100 g |
Each for $100.89
|
|
|||||
AAA1426336
|
Thermo Scientific Chemicals
A1426336 |
500 g |
Each for $312.74
|
|
|||||
Description
Phenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylacetaldehyde is used for the preparation of fragrances and polymers like polyesters, which find application as a rate controlling additive in polymerization reactions. It is an active component of fragrances and floral scent due to its honey-like sweet character and finds application in flavored cigarettes and beverages. It is an insect attractant and utilized in blacklight trap for pests. It is also used as a building block in the synthesis of more complex chemicals, such as resmethrin. Furthermore, it is used in association with acetic anhydride and allyltrimethylsilane in three-component coupling process catalyzed by LiBF4 providing homoallylic acetates.
Solubility
Miscible with alcohol and diethyl ether. Slightly miscible with water.
Notes
Store in cool place. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
122-78-1 | |
120.15 | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
998 | |
2-phenylacetaldehyde |
C8H8O | |
MFCD00006993 | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
CHEBI:16424 | |
O=CCC1=CC=CC=C1 |
Specifications
122-78-1 | |
194°C to 196°C | |
C8H8O | |
MFCD00006993 | |
385791 | |
14,7265 | |
Miscible with alcohol and diethyl ether. Slightly miscible with water. | |
O=CCC1=CC=CC=C1 | |
120.15 | |
CHEBI:16424 | |
95% |
1.075 | |
68°C (154°F) | |
1.535 | |
25 g | |
Air sensitive | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
2-phenylacetaldehyde | |
998 | |
120.15 | |
Phenylacetaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P330-P333+P313-P363-P370+P378q-P501c
H227-H302-H317
EINECSNumber : 204-574-5
RTECSNumber : CY1420000
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only