Learn More
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 370911000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenylacetaldehyde | |
122-78-1 | |
Colorless to Yellow | |
193.0°C | |
Authentic | |
Glass bottle | |
1.5250 to 1.5320 | |
MFCD00006993 | |
07, 292 | |
15, 7377 | |
Solubility in water: 2.210g/L water (25°C). Other solubilities: soluble in ethanol,acetone,diethyl ether | |
O=CCC1=CC=CC=C1 | |
120.15 | |
998 | |
120.15 | |
Liquid |
98% | |
5mg KOH/g max. | |
1.0290g/mL | |
68°C | |
97.5% min. (GC) | |
C8H8O | |
C6H5CH2CHO | |
100 g | |
1.029 | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
2-phenylacetaldehyde | |
approx. 0.01% citric acid | |
CHEBI:16424 | |
98% |
Chemical Identifiers
122-78-1 | |
120.15 | |
DTUQWGWMVIHBKE-UHFFFAOYSA-N | |
998 | |
2-phenylacetaldehyde |
C8H8O | |
MFCD00006993 | |
phenylacetaldehyde, benzeneacetaldehyde, hyacinthin, alpha-tolualdehyde, phenylethanal, 2-phenylethanal, phenylacetic aldehyde, alpha-toluic aldehyde, acetaldehyde, phenyl, benzylcarboxaldehyde | |
CHEBI:16424 | |
O=CCC1=CC=CC=C1 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If s
GHS Signal Word: Warning
EINECSNumber : 204-574-5
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only