missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylacetyl chloride, 98%, Thermo Scientific Chemicals

$94.62 - $94.62

Chemical Identifiers

CAS 103-80-0
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000729
InChI Key VMZCDNSFRSVYKQ-UHFFFAOYSA-N
Synonym phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
PubChem CID 7679
IUPAC Name 2-phenylacetyl chloride
SMILES ClC(=O)CC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC130301000
SDS View Documents
Thermo Scientific Chemicals
130301000
100 g
Each for $94.62
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-80-0
154.59
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
7679
ClC(=O)CC1=CC=CC=C1
C8H7ClO
MFCD00000729
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
2-phenylacetyl chloride

Specifications

103-80-0
94.0°C to 95.0°C (12.0 mmHg)
97.5% min. (GC)
C8H7ClO
C6H5CH2COCl
100 g
1.16
Solubility in water: reacts with water
ClC(=O)CC1=CC=CC=C1
154.59
154.6
Phenylacetyl chloride, 98%
1.1600g/mL
102°C
Glass bottle
1.5320 to 1.5330
MFCD00000729
09,436
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
2-phenylacetyl chloride
7679
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-146-5

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only