missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylarsine Oxide Dechlorinating Agent, 1.2 g/L, Ricca Chemical
Supplier: Ricca Chemical Company 574516
Specifications
| Phenylarsine Oxide Dechlorinating Agent | |
| 0.13,97.0,0.45 | |
| C6H5AsO | |
| phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
| O=[As]C1=CC=CC=C1 | |
| 168.03 | |
| CHEBI:75253 | |
| 500 mL |
| 637-03-6,7732-18-5,1310-73-2 | |
| 0.13,100.0,0.47 | |
| MFCD00001990 | |
| BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
| arsorosobenzene | |
| 4778 | |
| Amber Glass Bottle | |
| 1.2g/L |
Chemical Identifiers
| 637-03-6,7732-18-5,1310-73-2 | |
| 0.13,97.0,0.45 | |
| 0.13,100.0,0.47 | |
| Phenylarsine Oxide Dechlorinating Agent |