missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Phenylarsine Oxide Solution, 0.00564N (Certified), Fisher Chemical™
Supplier: Fisher Chemical SP681
Specifications
Phenylarsine Oxide Solution | |
0.00564N | |
100°C | |
C6H5AsO | |
0.00559 to 0.00569N | |
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
O=[As]C1=CC=CC=C1 | |
168.03 | |
CHEBI:75253 | |
0.00564N | |
Glass Bottle | |
1 L |
Undesignated | |
637-03-6,1310-73-2,7732-18-5 | |
0°C | |
MFCD00001990 | |
1.0g/cm³ | |
BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
arsorosobenzene | |
4778 | |
Certified | |
Pass Test | |
6.5 | |
Liquid |
Chemical Identifiers
637-03-6 | |
168.03 | |
BQVCCPGCDUSGOE-UHFFFAOYSA-N | |
4778 | |
arsorosobenzene |
C6H5AsO | |
MFCD00001990 | |
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060 | |
CHEBI:75253 | |
O=[As]C1=CC=CC=C1 |
Safety and Handling
