missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylarsine Oxide Solution, 0.00564N (Certified), Fisher Chemical™

Supplier:  Thermo Fisher Scientific SP684

 View more versions of this product

Catalog No. SP68-4



Explore available promotions

Description

Description

Specifications

Specifications

Phenylarsine Oxide Solution
0.00564N
100°C
C6H5AsO
0.00559 to 0.00569N
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
O=[As]C1=CC=CC=C1
168.03
CHEBI:75253
0.00564N
Glass Bottle
4 L
Undesignated
637-03-6,1310-73-2,7732-18-5
0°C
MFCD00001990
1.0g/cm³
BQVCCPGCDUSGOE-UHFFFAOYSA-N
arsorosobenzene
4778
Certified
Pass Test
6.5
Liquid

Chemical Identifiers

637-03-6
168.03
BQVCCPGCDUSGOE-UHFFFAOYSA-N
4778
arsorosobenzene
C6H5AsO
MFCD00001990
phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
CHEBI:75253
O=[As]C1=CC=CC=C1
Safety and Handling

Safety and Handling

SDS Documents

Documents

Accessories
Promotions

Promotions