missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylethyl Alcohol, USP, Spectrum™ Chemical
P1058, 60-12-8, C8H10O
$453.71 - $51470.49
Chemical Identifiers
CAS | 60-12-8 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002886 |
InChI Key | WRMNZCZEMHIOCP-UHFFFAOYSA-N |
IUPAC Name | 2-phenylethan-1-ol |
SMILES | OCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18605137
|
Spectrum Chemical Mfg Cor
P1058100GM |
100 g | Amber Glass Bottle |
Each for $453.71
|
|
|||||
18605140
|
Spectrum Chemical Mfg Cor
P1058200LTBL |
200 L | Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
18605138
|
Spectrum Chemical Mfg Cor
P10581KG |
1 kg | Amber Glass Bottle |
Each for $1,030.49
|
|
|||||
18605139
|
Spectrum Chemical Mfg Cor
P1058159KGBL |
15.9 kg | Steel Pail |
Each for $6,048.23
|
|
|||||
Description
Spectrum™ Chemical Phenylethyl Alcohol, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
60-12-8 | |
122.17 | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
OCCC1=CC=CC=C1 |
C8H10O | |
MFCD00002886 | |
2-phenylethan-1-ol |
Specifications
60-12-8 | |
5e-005 | |
C8H10O | |
MFCD00002886 | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
2-phenylethan-1-ol | |
USP |
1 | |
Amber Glass Bottle | |
1.531-1.534 | |
100 g | |
OCCC1=CC=CC=C1 | |
122.17 |