Learn More
Phenylethyl isothiocyanate, 96%, Thermo Scientific Chemicals
$74.75 - $304.29
Chemical Identifiers
CAS | 2257-09-2 |
---|---|
Molecular Formula | C9H9NS |
Molecular Weight (g/mol) | 163.24 |
MDL Number | MFCD00004821 |
InChI Key | IZJDOKYDEWTZSO-UHFFFAOYSA-N |
ChEBI | CHEBI:351346 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC265160050
|
Thermo Scientific Chemicals
265160050 |
5 g | Glass bottle |
Each for $74.75
|
|
|||||
AC265160250
|
Thermo Scientific Chemicals
265160250 |
25 g | Glass bottle |
Each for $304.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2257-09-2 | |
163.24 | |
IZJDOKYDEWTZSO-UHFFFAOYSA-N |
C9H9NS | |
MFCD00004821 | |
CHEBI:351346 |
Specifications
2257-09-2 | |
1.1000g/mL | |
100°C | |
96% | |
C9H9NS | |
C6H5CH2CH2NCS | |
5 g | |
1.1 | |
2,6-dichlorofluorobenzene, 1,3-dichloro-2-fluoro-benzene, 2,6-dichloro-1-fluorobenzene, 2,6-dichloro fluoro benzene, benzene,1,3-dichloro-2-fluoro, benzene, 1,3-dichloro-2-fluoro, pubchem2311, 2,6-dichlorfluorbenzol, acmc-2097ae, 2,6-dichloro fluorobenzene | |
IZJDOKYDEWTZSO-UHFFFAOYSA-N | |
1,3-dichloro-2-fluorobenzene | |
137520 | |
96% | |
Phenylethyl isothiocyanate, 96% |
Colorless to Yellow | |
113.0°C (1.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5860 to 1.5910 | |
MFCD00004821 | |
12, 1100 | |
15, 7340 | |
Solubility in water: insoluble | |
S=C=NCCC1=CC=CC=C1 | |
163.24 | |
163.24 | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT i
GHS Signal Word: Danger
EINECSNumber : 218-855-5
RTECSNumber : NX9115000
TSCA : TSCA
RUO – Research Use Only