Learn More
Phenylhydrazine Hydrochloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 151551000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenylhydrazine hydrochloride | |
59-88-1 | |
Beige-Pink or White to Yellow | |
99% min. (HPLC) | |
C6H9ClN2 | |
MFCD00012924 | |
15, 67 | |
15, 7404 | |
(5% in H2O) Clear colorless to light yellow solution | |
[H+].[Cl-].NNC1=CC=CC=C1 | |
144.60 | |
144.61 | |
Fine Crystalline Powder |
99+% | |
250.0°C to 254.0°C | |
Authentic | |
Plastic bottle | |
C6H5NHNH2·HCl | |
100 g | |
07,284 | |
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707 | |
JOVOSQBPPZZESK-UHFFFAOYSA-N | |
phenylhydrazine;hydrochloride | |
60962 | |
99+% |
Chemical Identifiers
59-88-1 | |
144.60 | |
JOVOSQBPPZZESK-UHFFFAOYSA-N | |
60962 | |
[H+].[Cl-].NNC1=CC=CC=C1 |
C6H9ClN2 | |
MFCD00012924 | |
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707 | |
phenylhydrazine;hydrochloride |
Safety and Handling
GHS H Statement
Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May cause cancer. May cause an allergic skin reaction. Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.
GHS Signal Word: Danger
EINECSNumber : 200-444-7
RTECSNumber : MV9000000
TSCA : TSCA
RUO – Research Use Only