missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylhydrazine Hydrochloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 151555000

 View more versions of this product

Catalog No. AC151555000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenylhydrazine hydrochloride
59-88-1
Beige-Pink or White to Yellow
99% min. (HPLC)
C6H9ClN2
MFCD00012924
15, 67
15, 7404
(5% in H2O) Clear colorless to light yellow solution
[H+].[Cl-].NNC1=CC=CC=C1
144.60
144.61
Fine Crystalline Powder
99+%
250.0°C to 254.0°C
Authentic
Plastic bottle
C6H5NHNH2·HCl
500 g
07,284
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
JOVOSQBPPZZESK-UHFFFAOYSA-N
phenylhydrazine;hydrochloride
60962
99+%

Chemical Identifiers

59-88-1
144.60
JOVOSQBPPZZESK-UHFFFAOYSA-N
60962
[H+].[Cl-].NNC1=CC=CC=C1
C6H9ClN2
MFCD00012924
phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
phenylhydrazine;hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled. Toxic if swallowed. Toxic in contact with skin. Suspected of causing genetic defects. May cause cancer. May cause an allergic skin reaction. Causes skin irritation. Causes serious eye irritation. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment.

GHS Signal Word: Danger

EINECSNumber : 200-444-7

RTECSNumber : MV9000000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only