Learn More
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals
$207.93 - $207.93
Chemical Identifiers
CAS | 100-58-3 |
---|---|
Molecular Formula | C6H5BrMg |
Molecular Weight (g/mol) | 181.32 |
MDL Number | MFCD00000038 |
InChI Key | ANRQGKOBLBYXFM-UHFFFAOYSA-M |
Synonym | phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, magnesium, bromophenyl, phenylmagnesiumbromide, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, phenyhnagnesium bromide |
PubChem CID | 66852 |
IUPAC Name | magnesium;benzene;bromide |
SMILES | Br[Mg]C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC445961000
|
Thermo Scientific Chemicals
445961000 |
100 mL | AcroSeal™ Glass bottle |
Each for $207.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-58-3 | |
181.32 | |
ANRQGKOBLBYXFM-UHFFFAOYSA-M | |
66852 | |
Br[Mg]C1=CC=CC=C1 |
C6H5BrMg | |
MFCD00000038 | |
phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, magnesium, bromophenyl, phenylmagnesiumbromide, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, phenyhnagnesium bromide | |
magnesium;benzene;bromide |
Specifications
100-58-3,5614-37-9 | |
AcroSeal™ Glass bottle | |
C6H5MgBr | |
100 mL | |
Solubility in water: reacts | |
Br[Mg]C1=CC=CC=C1 | |
181.32 | |
181.31 |
−1°C | |
C6H5BrMg | |
MFCD00000038 | |
phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, magnesium, bromophenyl, phenylmagnesiumbromide, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, phenyhnagnesium bromide | |
ANRQGKOBLBYXFM-UHFFFAOYSA-M | |
magnesium;benzene;bromide | |
66852 | |
Phenylmagnesium bromide |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 202-867-2
RUO – Research Use Only