missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylmethylsulfonyl Fluoride Crystalline MP Biomedicals
$98.79 - $1606.67
Chemical Identifiers
CAS | 329-98-6 |
---|---|
Molecular Formula | C7H7FO2S |
Molecular Weight (g/mol) | 174.189 |
InChI Key | YBYRMVIVWMBXKQ-UHFFFAOYSA-N |
Synonym | phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride |
PubChem CID | 4784 |
ChEBI | CHEBI:8102 |
IUPAC Name | phenylmethanesulfonyl fluoride |
SMILES | C1=CC=C(C=C1)CS(=O)(=O)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN19538101
|
MP Biomedicals Inc
0219538101 |
1 g |
Each for $98.79
|
|
|||||
ICN19538105
|
MP Biomedicals Inc
0219538105 |
5 g |
Each for $148.38
|
|
|||||
ICN19538125
|
MP Biomedicals Inc
0219538125 |
25 g |
Each for $482.38
|
|
|||||
ICN19538180
|
MP Biomedicals Inc
0219538180 |
100 g |
Each for $1,606.67
|
|
|||||
Chemical Identifiers
329-98-6 | |
174.189 | |
phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
CHEBI:8102 | |
C1=CC=C(C=C1)CS(=O)(=O)F |
C7H7FO2S | |
YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
4784 | |
phenylmethanesulfonyl fluoride |
Specifications
329-98-6 | |
1 g | |
YBYRMVIVWMBXKQ-UHFFFAOYSA-N | |
phenylmethanesulfonyl fluoride | |
4784 | |
174.2 | |
Phenylmethylsulfonyl Fluoride |
C7H7FO2S | |
phenylmethylsulfonyl fluoride, pmsf, benzenemethanesulfonyl fluoride, benzylsulfonyl fluoride, alpha-toluenesulfonyl fluoride, benzylsulphonyl fluoride, alpha-toluenesulphonyl fluoride, phenylmethylsulfonylfluoride, phenylmethylsulphonyl fluoride, .alpha.-toluenesulfonyl fluoride | |
C1=CC=C(C=C1)CS(=O)(=O)F | |
174.189 | |
CHEBI:8102 | |
Crystalline |