Learn More
Phenylpropiolic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1516309
Description
Phenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenylpropiolic acid is used as a Pharmaceutical Intermediates, laboratory chemicals.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Phenylpropiolic acid | |
135°C to 138°C | |
MFCD00004361 | |
742587 | |
Soluble in water. | |
OC(=O)C#CC1=CC=CC=C1 | |
146.15 | |
146.15 |
637-44-5 | |
C9H6O2 | |
10 g | |
phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid | |
XNERWVPQCYSMLC-UHFFFAOYSA-N | |
3-phenylprop-2-ynoic acid | |
69475 | |
≥98% |
Chemical Identifiers
637-44-5 | |
146.15 | |
XNERWVPQCYSMLC-UHFFFAOYSA-N | |
69475 | |
OC(=O)C#CC1=CC=CC=C1 |
C9H6O2 | |
MFCD00004361 | |
phenylpropiolic acid, 3-phenylpropiolic acid, phenylpropynoic acid, 3-phenylpropynoic acid, 2-propynoic acid, 3-phenyl, phenylacetylenecarboxylic acid, 3-phenyl-2-propynoic acid, phenylacetylene monocarboxylic acid, propiolic acid, 3-phenyl, phenyl propiolic acid | |
3-phenylprop-2-ynoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-285-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only