missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylselenyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 179510100

 View more versions of this product

Catalog No. AC179510100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

Phenylselenyl chloride
5707-04-0
Orange
Authentic
Glass bottle
C6H5SeCl
10 g
05,518; 06,459; 07,286; 09,25; 11,33; 13,26; 16,19; 17,26
WJCXADMLESSGRI-UHFFFAOYSA-N
phenyl selenohypochlorite
21928
98%
98%
61.0°C to 65.0°C
120.0°C (20.0 mmHg)
97.5% min. (Argentometry)
C6H5ClSe
MFCD00000478
06, III, 1110
phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
Cl[Se]C1=CC=CC=C1
191.53
191.52
Crystalline Powder or Crystals

Chemical Identifiers

5707-04-0
191.53
WJCXADMLESSGRI-UHFFFAOYSA-N
21928
Cl[Se]C1=CC=CC=C1
C6H5ClSe
MFCD00000478
phenylselenenyl chloride, phenylselenyl chloride, benzeneselenenyl chloride, chloroselenobenzene, phenyl hypochloroselenoite, phenylselenium chloride, phsecl, phenylselenenylchloride, phenylselanyl chlorane, chloroselanylbenzene
phenyl selenohypochlorite
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 227-196-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only