missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-Phenylsuccinic acid, 98%, Thermo Scientific Chemicals

$53.33 - $693.95

Chemical Identifiers

CAS 635-51-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00004256
InChI Key LVFFZQQWIZURIO-UHFFFAOYSA-N
Synonym phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
PubChem CID 95459
IUPAC Name 2-phenylbutanedioic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1197114
View Documents
Thermo Scientific Chemicals
A1197114
25 g
Each for $53.33
 
AAA1197122
View Documents
Thermo Scientific Chemicals
A1197122
100 g
Each for $153.58
 
AAA1197136
View Documents
Thermo Scientific Chemicals
A1197136
500 g
Each for $693.95
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

635-51-8
194.186
LVFFZQQWIZURIO-UHFFFAOYSA-N
95459
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
C10H10O4
MFCD00004256
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
2-phenylbutanedioic acid

Specifications

635-51-8
C10H10O4
25 g
phenylsuccinic acid, dl-phenylsuccinic acid, 2-phenylsuccinic acid, phenyl succinic acid, butanedioic acid, phenyl, succinic acid, phenyl, 2-phenylsuccinate, .alpha.-phenylsuccinic acid, phenylsuccinate, butanedioic acid, 2-phenyl
C1=CC=C(C=C1)C(CC(=O)O)C(=O)O
194.186
194.19
(±)-Phenylsuccinic acid
167°C to 169°C
MFCD00004256
1875051
LVFFZQQWIZURIO-UHFFFAOYSA-N
2-phenylbutanedioic acid
95459
98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-238-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only