missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylthiohydantoin-tyrosine 98.0+%, TCI America™
$87.60 - $329.94
Chemical Identifiers
| CAS | 4332-95-0 |
|---|---|
| Molecular Formula | C16H14N2O2S |
| Molecular Weight (g/mol) | 298.36 |
| MDL Number | MFCD00027415 |
| InChI Key | LCFDDJINTNHAEQ-UHFFFAOYSA-N |
| Synonym | 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, PTH-tyrosine |
| PubChem CID | 2872399 |
| IUPAC Name | 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one |
| SMILES | C1=CC=C(C=C1)N2C(=O)C(NC2=S)CC3=CC=C(C=C3)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P0368100MG
|
TCI America
P0368100MG |
100 mg |
Each for $87.60
|
|
|||||
|
P03681G
|
TCI America
P03681G |
1 g |
Each for $329.94
|
|
|||||
Chemical Identifiers
| 4332-95-0 | |
| 298.36 | |
| LCFDDJINTNHAEQ-UHFFFAOYSA-N | |
| 2872399 | |
| C1=CC=C(C=C1)N2C(=O)C(NC2=S)CC3=CC=C(C=C3)O |
| C16H14N2O2S | |
| MFCD00027415 | |
| 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, PTH-tyrosine | |
| 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one |
Specifications
| 221°C | |
| White-Yellow | |
| MFCD00027415 | |
| LCFDDJINTNHAEQ-UHFFFAOYSA-N | |
| 5-[(4-hydroxyphenyl)methyl]-3-phenyl-2-sulfanylideneimidazolidin-4-one | |
| 298.36 | |
| 298.36 | |
| Crystalline Powder |
| 4332-95-0 | |
| C16H14N2O2S | |
| 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, PTH-tyrosine | |
| C1=CC=C(C=C1)N2C(=O)C(NC2=S)CC3=CC=C(C=C3)O | |
| 100 mg | |
| 2872399 | |
| ≥98.0% (HPLC) | |
| Phenylthiohydantoin-tyrosine |
Safety and Handling
RTECSNumber : NJ0730000
TSCA : No