missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylthiophosphonic Dichloride 98.0+%, TCI America™
$85.75 - $508.33
Chemical Identifiers
| CAS | 3497-00-5 |
|---|---|
| Molecular Formula | C6H5Cl2PS |
| Molecular Weight (g/mol) | 211.04 |
| MDL Number | MFCD00004916 |
| InChI Key | SXIWNIQDOJKDGB-UHFFFAOYSA-N |
| Synonym | Phenylphosphonothioic Dichloride |
| PubChem CID | 19016 |
| IUPAC Name | dichloro-phenyl-sulfanylidene-$l^{5}-phosphane |
| SMILES | C1=CC=C(C=C1)P(=S)(Cl)Cl |
Chemical Identifiers
| 3497-00-5 | |
| 211.04 | |
| SXIWNIQDOJKDGB-UHFFFAOYSA-N | |
| 19016 | |
| C1=CC=C(C=C1)P(=S)(Cl)Cl |
| C6H5Cl2PS | |
| MFCD00004916 | |
| Phenylphosphonothioic Dichloride | |
| dichloro-phenyl-sulfanylidene-$l^{5}-phosphane |
Specifications
| 3497-00-5 | |
| 270°C | |
| MFCD00004916 | |
| 3265 | |
| SXIWNIQDOJKDGB-UHFFFAOYSA-N | |
| dichloro-phenyl-sulfanylidene-$l^{5}-phosphane | |
| 19016 | |
| ≥98.0% (GC,T) | |
| Phenylthiophosphonic Dichloride |
| Yellow | |
| C6H5Cl2PS | |
| 25 g | |
| Phenylphosphonothioic Dichloride | |
| C1=CC=C(C=C1)P(=S)(Cl)Cl | |
| 211.04 | |
| 211.04 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2511&(3)-2612
TSCA : Yes