missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2285122

 View more versions of this product

Catalog No. AAB2285122


Explore available promotions

Description

Description

Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

Solubility
Soluble in water 333 g/L.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
Specifications

Specifications

Phenyltrimethylammonium chloride
∼236°C (sublimation)
MFCD00011790
UN2811
Hygroscopic
Soluble in water 333g/L.
[Cl-].C[N+](C)(C)C1=CC=CC=C1
171.67
171.67
138-24-9
C9H14ClN
100 g
3572527
phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
MQAYPFVXSPHGJM-UHFFFAOYSA-M
N,N,N-trimethylanilinium chloride
67309
≥98%

Chemical Identifiers

138-24-9
171.67
MQAYPFVXSPHGJM-UHFFFAOYSA-M
67309
C9H14ClN
MFCD00011790
phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
[Cl-].C[N+](C)(C)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c

H301+H311

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-319-0

RTECSNumber : BT2190000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only