missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium iodide, 99%, Thermo Scientific Chemicals

$137.33 - $573.42

Chemical Identifiers

CAS 98-04-4
Molecular Formula C9H14IN
Molecular Weight (g/mol) 263.122
MDL Number MFCD00011791
InChI Key KKLAORVGAKUOPZ-UHFFFAOYSA-M
Synonym phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
PubChem CID 94135
IUPAC Name trimethyl(phenyl)azanium;iodide
SMILES C[N+](C)(C)C1=CC=CC=C1.[I-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1440222
View Documents
Thermo Scientific Chemicals
A1440222
100 g
Each for $137.33
 
AAA1440236
View Documents
Thermo Scientific Chemicals
A1440236
500 g
Each for $573.42
 
Description

Description

Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium iodide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in water and methanol.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

98-04-4
263.122
KKLAORVGAKUOPZ-UHFFFAOYSA-M
94135
C[N+](C)(C)C1=CC=CC=C1.[I-]
C9H14IN
MFCD00011791
phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
trimethyl(phenyl)azanium;iodide

Specifications

98-04-4
C9H14IN
100 g
3916752
14,7318
Soluble in water and methanol.
C[N+](C)(C)C1=CC=CC=C1.[I-]
263.122
263.12
Phenyltrimethylammonium iodide
∼227°C (sublimation)
MFCD00011791
UN2811
Light sensitive
phenyltrimethylammonium iodide, trimethylphenylammonium iodide, n,n,n-trimethylanilinium iodide, trimethylanilinium iodide, trimethylanillinium iodide, n,n-dimethylaniline methiodide, unii-4o888xq7fp, ammonium, phenyltrimethyl-, iodide, n,n,n-trimethylbenzenaminium iodide, benzenaminium, n,n,n-trimethyl-, iodide
KKLAORVGAKUOPZ-UHFFFAOYSA-M
trimethyl(phenyl)azanium;iodide
94135
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c

H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-630-3

RTECSNumber : BT2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only