missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylurea, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 131021000

 View more versions of this product

Catalog No. AC131021000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Phenylurea
64-10-8
Authentic
Plastic bottle
C6H5NHCONH2
100g
15, 7429
Solubility in water: 1g/100mL. Other solubilities: soluble in hot alcohol,ether,ethylacetate,and glacial acetic acid
C1=CC=C(C=C1)NC(=O)N
136.15
136.15
97%
238°C
96% min. (HPLC)
C7H8N2O
MFCD00007944
12, 346
1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea
LUBJCRLGQSPQNN-UHFFFAOYSA-N
phenylurea
6145
97%

Chemical Identifiers

64-10-8
136.15
LUBJCRLGQSPQNN-UHFFFAOYSA-N
6145
C1=CC=C(C=C1)NC(=O)N
C7H8N2O
MFCD00007944
1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea
phenylurea
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

GHS Signal Word: Warning

EINECSNumber : 200-576-5

RTECSNumber : YU0650000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only