missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylurea, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131022500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phenylurea | |
64-10-8 | |
Authentic | |
Plastic bottle | |
C6H5NHCONH2 | |
250g | |
15, 7429 | |
Solubility in water: 1g/100mL. Other solubilities: soluble in hot alcohol,ether,ethylacetate,and glacial acetic acid | |
C1=CC=C(C=C1)NC(=O)N | |
136.15 | |
136.15 |
97% | |
238°C | |
96% min. (HPLC) | |
C7H8N2O | |
MFCD00007944 | |
12, 346 | |
1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea | |
LUBJCRLGQSPQNN-UHFFFAOYSA-N | |
phenylurea | |
6145 | |
97% |
Chemical Identifiers
64-10-8 | |
136.15 | |
LUBJCRLGQSPQNN-UHFFFAOYSA-N | |
6145 | |
C1=CC=C(C=C1)NC(=O)N |
C7H8N2O | |
MFCD00007944 | |
1-phenylurea, n-phenylurea, urea, phenyl, phenylcarbamide, monophenylurea, stabilizer vh, stabilisator vh, phenyl-urea, urea, n-phenyl, phenyl urea | |
phenylurea |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
GHS Signal Word: Warning
EINECSNumber : 200-576-5
RTECSNumber : YU0650000
TSCA : TSCA
RUO â Research Use Only